siRNA Details
Gene Symbol: HECW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287059.1 | NP_001273988.1 | 6496 | |||
NM_015052.4 | NP_055867.3 | 6833 | ||||
GenBank mRNA | AB048365.1 | - | 6184 | |||
BC151227.1 | - | 6832 | 9007236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
