siRNA Details
Gene Symbol: PIGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002642.3 | NP_002633.1 | 2 | 1223 | ||
NM_153747.1 | NP_714969.1 | 2 | 1200 | |||
GenBank mRNA | AK308201.1 | - | 2 | 1180 | ||
BC006539.2 | - | 2 | 1170 | 2823399 | ||
D85418.1 | - | 1 | 1182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
