siRNA Details
Gene Symbol: ATP2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021949.3 | NP_068768.2 | 20 | 3563 | ||
XM_005274692.3 | XP_005274749.1 | 22 | 3871 | |||
GenBank mRNA | AK090838.1 | - | 1 | 1804 | ||
AK307765.1 | - | 4 | 489 | |||
BM695792.1 | - | 3 | 383 | |||
U15689.1 | - | 172 | ||||
U15690.1 | - | 1 | 104 | |||
U57971.1 | - | 20 | 3563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
