siRNA Details
Gene Symbol: HDAC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159286.1 | NP_001152758.1 | 19 | 2342 | ||
NM_032019.5 | NP_114408.3 | 20 | 2402 | |||
GenBank mRNA | AF426160.1 | - | 20 | 2382 | ||
BC032049.1 | - | 20 | 2215 | |||
BC094734.1 | - | 18 | 2070 | |||
BC125082.1 | - | 14 | 1551 | |||
BC125083.1 | - | 20 | 2076 | 40117772 | ||
CR456465.1 | - | 20 | 2085 |
