siRNA Details
Gene Symbol: BCL2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191.3 | NP_001182.1 | 3 | 1390 | ||
NM_001317919.1 | NP_001304848.1 | 3 | 1561 | |||
NM_001317920.1 | NP_001304849.1 | 3 | 1216 | |||
NM_001317921.1 | NP_001304850.1 | 3 | 1345 | |||
NM_001322239.1 | NP_001309168.1 | 3 | 1234 | |||
NM_001322240.1 | NP_001309169.1 | 3 | 1363 | |||
NM_001322242.1 | NP_001309171.1 | 3 | 1216 | |||
NM_138578.2 | NP_612815.1 | 3 | 1579 | |||
NR_134257.1 | 3 | 1237 | ||||
XM_011528964.2 | XP_011527266.1 | 3 | 1198 | |||
XM_017027993.1 | XP_016883482.1 | 3 | 1204 | |||
GenBank mRNA | BC019307.1 | - | 3 | 1334 | 2823498 | |
BX647525.1 | - | 5 | 1115 | |||
CR936637.1 | - | 3 | 1365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
