siRNA Details
Gene Symbol: ATP1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144699.3 | NP_653300.2 | 3 | 711 | ||
XM_011509582.1 | XP_011507884.1 | 2 | 178 | |||
GenBank mRNA | AF506797.1 | - | 3 | 703 | ||
AK308286.1 | - | 3 | 669 | |||
AL833335.1 | - | 3 | 684 | |||
BC094801.1 | - | 3 | 662 | 5268907 | ||
BX647234.1 | - | 3 | 685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
