siRNA Details
Gene Symbol: DIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145173.3 | NP_660156.1 | 2 | 2084 | ||
XM_011527713.2 | XP_011526015.1 | 2 | 2249 | |||
GenBank mRNA | AK124445.1 | - | 1 | 1395 | ||
AK124672.1 | - | 1 | 848 | |||
BC030660.1 | - | 2 | 2063 | 4814337 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
