siRNA Details
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006638.2 | NP_001006639.1 | 9 | 2187 | ||
NM_001163989.1 | NP_001157461.1 | 9 | 2336 | |||
NM_001163990.1 | NP_001157462.1 | 8 | 2076 | |||
NM_175738.4 | NP_783865.1 | 9 | 2580 | |||
XM_005257288.4 | XP_005257345.1 | 9 | 2205 | |||
GenBank mRNA | AK057069.1 | - | 3 | 1731 | ||
AK127073.1 | - | 8 | 3340 | |||
BC016615.1 | - | 10 | 1189 | 4520191 | ||
BC040547.1 | - | 10 | 1265 | 5287038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
