siRNA Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001196.3 | NP_001187.1 | 6 | 1484 | ||
NM_001244567.1 | NP_001231496.1 | 6 | 1395 | |||
NM_001244569.1 | NP_001231498.1 | 5 | 1273 | |||
NM_001244570.1 | NP_001231499.1 | 4 | 1203 | |||
NM_001244572.1 | NP_001231501.1 | 4 | 1521 | |||
NM_197966.2 | NP_932070.1 | 6 | 1802 | |||
NM_197967.2 | NP_932071.1 | 5 | 1414 | |||
GenBank mRNA | AK094795.1 | - | 6 | 1743 | ||
BC036364.1 | - | 6 | 1781 | 4819949 | ||
D89803.1 | - | 5 | 1383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
