siRNA Details
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002687.3 | NP_002678.2 | 9 | 2318 | ||
GenBank mRNA | AK223612.1 | - | 9 | 2253 | ||
AK292579.1 | - | 9 | 2302 | |||
AK300164.1 | - | 2 | 1567 | |||
AK303136.1 | - | 5 | 1909 | |||
AK304622.1 | - | 1 | 1423 | |||
BC062602.1 | - | 9 | 2229 | 5491466 | ||
U77718.1 | - | 9 | 2329 | |||
Y09703.1 | - | 9 | 2234 |
