siRNA Details
Gene Symbol: ATP5J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003713.2 | NP_001003713.1 | 3 | 212 | ||
NM_004889.3 | NP_004880.1 | 3 | 230 | |||
GenBank mRNA | AF047436.1 | - | 3 | 197 | ||
AF088918.1 | - | 3 | 183 | |||
AK094388.1 | - | 1 | 593 | |||
AK123419.1 | - | 1 | 2256 | |||
AK223348.1 | - | 3 | 193 | |||
AY046911.1 | - | 3 | 172 | |||
BC003678.1 | - | 3 | 193 | 3826469 | ||
BU587946.1 | - | 3 | 200 | |||
CB155273.1 | - | 3 | 197 | |||
CD173604.1 | - | 3 | 234 | |||
CR456891.1 | - | 3 | 170 | |||
CR542155.1 | - | 3 | 170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
