siRNA Details
Gene Symbol: TUBA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026516.1 | - | 4 | 1514 | ||
AK026632.1 | - | 4 | 1514 | |||
AK056693.1 | - | 3 | 2422 | |||
AK057773.1 | - | 4 | 1514 | |||
AK094717.1 | - | 3 | 3087 | |||
AK098771.1 | - | 4 | 1514 | |||
AK289354.1 | - | 4 | 1514 | |||
AK293337.1 | - | 2 | 1139 | |||
BC009512.2 | - | 4 | 1498 | 3537301 | ||
BC017004.1 | - | 4 | 1470 | 4430457 | ||
BC030820.1 | - | 4 | 1511 | 4277263 | ||
K00558.1 | - | 4 | 1486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
