siRNA Details
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000135.2 | NP_000126.2 | 22, 23 | 2057 | ||
NM_001286167.1 | NP_001273096.1 | 22, 23 | 2057 | |||
XM_005256294.4 | XP_005256351.1 | 22, 23 | 2058 | |||
XM_011522945.2 | XP_011521247.1 | 22, 23 | 2058 | |||
XM_011522946.2 | XP_011521248.1 | 22, 23 | 2065 | |||
XM_011522947.2 | XP_011521249.1 | 16, 17 | 1976 | |||
XM_011522948.2 | XP_011521250.1 | 22, 23 | 2058 | |||
XM_017023044.1 | XP_016878533.1 | 22, 23 | 2058 | |||
XM_017023045.1 | XP_016878534.1 | 22, 23 | 2058 | |||
GenBank mRNA | AK299282.1 | - | 22, 23 | 2046 | ||
X99226.1 | - | 22, 23 | 2046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
