siRNA Details
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 7, 8 | 648 | ||
XM_011522928.1 | XP_011521230.1 | 7, 8 | 826 | |||
XM_011522929.1 | XP_011521231.1 | 7, 8 | 691 | |||
XM_011522930.2 | XP_011521232.1 | 7, 8 | 659 | |||
XM_011522931.2 | XP_011521233.1 | 7, 8 | 659 | |||
XM_017023012.1 | XP_016878501.1 | 7, 8 | 675 | |||
XM_017023014.1 | XP_016878503.1 | 3, 4 | 775 | |||
GenBank mRNA | AJ441184.1 | - | 7, 8 | 647 | ||
AK056456.1 | - | 7, 8 | 647 | |||
AK074168.1 | - | 6, 7 | 1065 | |||
AK128387.1 | - | 7, 8 | 744 | |||
AK315590.1 | - | 7, 8 | 651 | |||
BC013735.1 | - | 5, 6 | 434 | |||
BC032542.1 | - | 7, 8 | 654 | 5534376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:

Citations (8)
The chromatin remodeling factor Mi-2alpha acts as a novel co-activator for human c-Myb.
- Authors
- Saether T, Berge T, Ledsaak M, Matre V, Alm-Kristiansen AH, Dahle O, Aubry F, Gabrielsen OS
- Journal
- J Biol Chem - Page No 13994 (Fri May 11 00:00:00 UTC 2007)
- Volume (Year)
- 282 (2007)
- Product Usage
- -
The estrogen and c-Myc target gene HSPC111 is over-expressed in breast cancer and associated with poor patient outcome.
- Authors
- Butt AJ, Sergio CM, Inman CK, Anderson LR, McNeil CM, Russell AJ, Nousch M, Preiss T, Biankin AV, Sutherland RL, Musgrove EA
- Journal
- Breast Cancer Res - Page No R28 (Tue Jan 01 00:00:00 UTC 2008)
- Volume (Year)
- 10 (2008)
- Product Usage
- -
PcG proteins, DNA methylation, and gene repression by chromatin looping.
- Authors
- Tiwari VK, McGarvey KM, Licchesi JD, Ohm JE, Herman JG, Schubeler D, Baylin SB
- Journal
- Plos Biol - Page No 2911 (Tue Dec 02 00:00:00 UTC 2008)
- Volume (Year)
- 6 (2008)
- Product Usage
- -
Genome-wide changes accompanying knockdown of fatty acid synthase in breast cancer.
- Authors
- Knowles LM, Smith JW
- Journal
- Bmc Genomics - Page No 168 (Mon Jan 01 00:00:00 UTC 2007)
- Volume (Year)
- 8 (2007)
- Product Usage
- -
Double PHD fingers protein DPF2 recognizes acetylated histones and suppresses the function of estrogen-related receptor alpha through histone deacetylase 1.
- Authors
- Matsuyama R, Takada I, Yokoyama A, Fujiyma-Nakamura S, Tsuji N, Kitagawa H, Fujiki R, Kim M, Kouzu-Fujita M, Yano T, Kato S
- Journal
- J Biol Chem - Page No 18166 (Fri Jun 11 00:00:00 UTC 2010)
- Volume (Year)
- 285 (2010)
- Product Usage
- -
Group VIA calcium-independent phospholipase A2 mediates endothelial cell S phase progression.
- Authors
- Herbert SP, Walker JH
- Journal
- J Biol Chem - Page No 35709 (Fri Nov 24 00:00:00 UTC 2006)
- Volume (Year)
- 281 (2006)
- Product Usage
- -
Regulator of G-protein signalling 2 mRNA is differentially expressed in mammary epithelial subpopulations and over-expressed in the majority of breast cancers.
- Authors
- Smalley MJ, Iravani M, Leao M, Grigoriadis A, Kendrick H, Dexter T, Fenwick K, Regan JL, Britt K, McDonald S, Lord CJ, Mackay A, Ashworth A
- Journal
- Breast Cancer Res - Page No R85 (Mon Jan 01 00:00:00 UTC 2007)
- Volume (Year)
- 9 (2007)
- Product Usage
- -
Distinct sequence elements of cyclin B1 promote localization to chromatin, centrosomes, and kinetochores during mitosis.
- Authors
- Bentley AM, Normand G, Hoyt J, King RW
- Journal
- Mol Biol Cell - Page No 4847 (Sat Dec 01 00:00:00 UTC 2007)
- Volume (Year)
- 18 (2007)
- Product Usage
- -
