siRNA Details
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008405.1 | - | 8 | 1566 | 4043484 | |
M33665.1 | - | 4 | 848 | |||
M94891.1 | - | 6 | 1555 | |||
U18468.1 | - | 6 | 1474 | |||
U18469.1 | - | 5 | 1217 | |||
X17097.1 | - | 6 | 1561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
