siRNA Details
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203287.1 | NP_976032.2 | 5 | 865 | ||
GenBank mRNA | BC020711.1 | - | 5 | 855 | 4732286 |
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002781.3 | NP_002772.3 | 6 | 1255 | ||
XM_017027003.1 | XP_016882492.1 | 6 | 1249 | |||
GenBank mRNA | BC012607.1 | - | 6 | 1222 | 4043912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
