siRNA Details
Gene Symbol: TAS1R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152228.2 | NP_689414.1 | 3 | 1158 | ||
XM_017002435.1 | XP_016857924.1 | 3 | 1180 | |||
XM_017002436.1 | XP_016857925.1 | 3 | 1180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
