siRNA Details
Gene Symbol: CDY2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004825.2 | NP_004816.1 | 1 | 783 | ||
GenBank mRNA | AF080598.1 | - | 1 | 783 | ||
AK292233.1 | - | 1 | 756 |
Gene Symbol: CDY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001722.1 | NP_001001722.1 | 1 | 782 | ||
GenBank mRNA | BC069087.1 | - | 1 | 782 | 7216901 | |
BC130426.1 | - | 1 | 551 | 40146457 | ||
BC130428.1 | - | 1 | 551 | 40146459 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
