siRNA Details
Gene Symbol: COX7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC100852.1 | - | 4 | 313 | 40002220 | |
BC100853.1 | - | 4 | 313 | 40002221 | ||
BC100854.2 | - | 4 | 313 | 40002222 | ||
BC133654.1 | - | 4 | 313 | 40002224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
