siRNA Details
Gene Symbol: NBN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002485.4 | NP_002476.2 | 2, 3 | 282 | ||
XM_011517046.1 | XP_011515348.1 | 2, 3 | 309 | |||
XM_017013461.1 | XP_016868950.1 | 2, 3 | 209 | |||
GenBank mRNA | AF051334.1 | - | 2, 3 | 223 | ||
AF058696.2 | - | 2, 3 | 224 | |||
AK001017.1 | - | 2, 3 | 278 | |||
AK223256.1 | - | 2, 3 | 226 | |||
AK289848.1 | - | 2, 3 | 278 | |||
AK312410.1 | - | 2, 3 | 284 | |||
BC005293.1 | - | 2, 3 | 204 | |||
BC016762.1 | - | 2, 3 | 196 | |||
BC136802.1 | - | 2, 3 | 241 | 9020792 | ||
BC136803.1 | - | 2, 3 | 241 | 9020793 | ||
BC144413.1 | - | 2, 3 | 241 | |||
BC146797.1 | - | 2, 3 | 234 | 8860301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
