siRNA Details
Gene Symbol: IP6K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242829.1 | NP_001229758.1 | 2, 3 | 884 | ||
NM_153273.3 | NP_695005.1 | 2, 3 | 546 | |||
GenBank mRNA | AK289772.1 | - | 1, 2 | 351 | ||
BC012944.1 | - | 1, 2 | 347 | 3872311 | ||
BG720032.1 | - | 2, 3 | 552 | |||
D87452.2 | - | 2, 3 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
