siRNA Details
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204466.1 | NP_001191395.1 | 13, 14 | 1741 | ||
NM_001204467.1 | NP_001191396.1 | 14, 15 | 1969 | |||
NM_001204468.1 | NP_001191397.1 | 14, 15 | 2003 | |||
NM_005676.4 | NP_005667.2 | 14, 15 | 1972 | |||
NM_152856.2 | NP_690595.1 | 13, 14 | 1738 | |||
XM_005272677.4 | XP_005272734.1 | 14, 15 | 2022 | |||
XM_005272678.4 | XP_005272735.1 | 13, 14 | 1556 | |||
XM_005272679.4 | XP_005272736.1 | 13, 14 | 1553 | |||
XM_006724563.1 | XP_006724626.1 | 10, 11 | 1189 | |||
XM_011543989.2 | XP_011542291.1 | 6, 7 | 913 | |||
XM_017029884.1 | XP_016885373.1 | 13, 14 | 1892 | |||
XM_017029885.1 | XP_016885374.1 | 13, 14 | 1664 | |||
GenBank mRNA | AK025854.1 | - | 2, 3 | 345 | ||
AK098115.1 | - | 2581 | ||||
AK292758.1 | - | 13, 14 | 1692 | |||
AL137421.1 | - | 15, 16 | 1861 | |||
BC000681.2 | - | 11, 12 | 1194 | |||
BC003089.1 | - | 13, 14 | 1677 | 3536058 | ||
BC004181.2 | - | 14, 15 | 1925 | 2961094 | ||
BC008733.2 | - | 14, 15 | 1957 | 3163064 | ||
BC024153.2 | - | 13, 14 | 1587 | 3845258 | ||
BX537969.1 | - | 14, 15 | 1954 | |||
D50912.1 | - | 14, 15 | 1820 | |||
KU178411.1 | - | 12, 13 | 1345 | |||
KU178412.1 | - | 13, 14 | 1579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
