siRNA Details
Gene Symbol: TFPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005246818.1 | XP_005246875.1 | 8 | 995 | ||
XM_005246819.1 | XP_005246876.1 | 7 | 743 | |||
XM_006712720.3 | XP_006712783.1 | 8 | 782 | |||
XM_011511707.2 | XP_011510009.1 | 9 | 1123 | |||
XM_011511708.1 | XP_011510010.1 | 8 | 1057 | |||
XM_011511709.2 | XP_011510011.1 | 8 | 825 | |||
GenBank mRNA | AB209866.1 | - | 6 | 7511 | ||
AK313562.1 | - | 7 | 907 | |||
J03225.1 | - | 7 | 801 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
