siRNA Details
Gene Symbol: RAB6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243719.1 | NP_001230648.1 | 3, 4 | 158 | ||
NM_002869.4 | NP_002860.2 | 3, 4 | 703 | |||
GenBank mRNA | AF091030.1 | - | 2, 3 | 110 | ||
AF119836.1 | - | 3, 4 | 251 | |||
AF130122.1 | - | 3, 4 | 192 | |||
AF198616.1 | - | 3, 4 | 182 | |||
AF498941.1 | - | 3, 4 | 182 | |||
AK057157.1 | - | 3, 4 | 668 | |||
AK223551.1 | - | 3, 4 | 621 | |||
AK301534.1 | - | 3, 4 | 161 | |||
BC003617.2 | - | 3, 4 | 599 | 3506585 | ||
BC068486.1 | - | 4, 5 | 644 | 30342562 | ||
CR541984.1 | - | 3, 4 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
