siRNA Details
Gene Symbol: SZT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015284.3 | NP_056099.3 | 31 | 4520 | ||
XM_005270686.3 | XP_005270743.1 | 33 | 4752 | |||
XM_006710501.3 | XP_006710564.1 | 32 | 4639 | |||
XM_011541106.2 | XP_011539408.1 | 32 | 4790 | |||
XM_011541107.2 | XP_011539409.1 | 29 | 4287 | |||
XM_017000819.1 | XP_016856308.1 | 33 | 4752 | |||
XM_017000820.1 | XP_016856309.1 | 32 | 4581 | |||
XM_017000821.1 | XP_016856310.1 | 29 | 4273 | |||
GenBank mRNA | AB007936.2 | - | 17 | 2372 | ||
BC151232.1 | - | 17 | 2372 | 8860124 |
