siRNA Details
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001481.2 | NP_001001481.2 | 3 | 200 | ||
NM_001271015.1 | NP_001257944.1 | 2 | 167 | |||
NM_018299.4 | NP_060769.4 | 2 | 167 | |||
NR_073119.1 | 2 | 167 | ||||
NR_073120.1 | 2 | 167 | ||||
NR_073121.1 | 2 | 167 | ||||
GenBank mRNA | AK001873.1 | - | 2 | 82 | ||
AK024050.1 | - | 3 | 165 | |||
AK225554.1 | - | 2 | 98 | |||
AK295792.1 | - | 2 | 167 | |||
AY948289.1 | - | 3 | 136 | |||
BC016326.1 | - | 2 | 121 | 4093131 | ||
BU155172.1 | - | 2 | 113 | |||
BX538145.1 | - | 2 | 90 | |||
BX648427.1 | - | 2 | 96 | |||
CR457275.1 | - | 1 | 79 | |||
DA352184.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
