siRNA Details
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001798.4 | NP_001789.2 | 2 | 416 | ||
NM_052827.3 | NP_439892.2 | 2 | 416 | |||
XM_011537732.1 | XP_011536034.1 | 2 | 416 | |||
GenBank mRNA | AB012305.1 | - | 2 | 178 | ||
AK291941.1 | - | 2 | 355 | |||
BC003065.2 | - | 2 | 336 | 3504348 | ||
BJ991087.1 | - | 2 | 413 | |||
BT006821.1 | - | 2 | 176 | |||
DA814453.1 | - | 2 | 416 | |||
M68520.1 | - | 2 | 366 | |||
X61622.1 | - | 2 | 176 | |||
X62071.1 | - | 2 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
