siRNA Details
Gene Symbol: CNDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289916.1 | - | 6 | 782 | ||
AK314572.1 | - | 6 | 818 | |||
AY358756.1 | - | 6 | 916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
