siRNA Details
Gene Symbol: RGS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002926.3 | NP_002917.1 | 2 | 1258 | ||
NM_198229.2 | NP_937872.1 | 2 | 1258 | |||
XM_006713905.1 | XP_006713968.1 | 2 | 577 | |||
XM_006713906.2 | XP_006713969.1 | 2 | 577 | |||
XM_017008529.1 | XP_016864018.1 | 2 | 1827 | |||
XM_017008530.1 | XP_016864019.1 | 2 | 577 | |||
GenBank mRNA | AF030111.1 | - | 2 | 1257 | ||
AF035152.1 | - | 1 | 408 | |||
AF464735.1 | - | 1 | 457 | |||
AF464736.1 | - | 1 | 457 | |||
AK293767.1 | - | 2 | 510 | |||
BC028154.1 | - | 2 | 520 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
