siRNA Details
Gene Symbol: ITLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080878.2 | NP_543154.1 | 6 | 676 | ||
XM_011509210.1 | XP_011507512.1 | 8 | 1153 | |||
XM_011509211.1 | XP_011507513.1 | 8 | 1150 | |||
GenBank mRNA | AY065973.1 | - | 6 | 648 | ||
AY358905.1 | - | 6 | 676 | |||
BC117225.1 | - | 6 | 674 | 40125776 | ||
BC143339.1 | - | 5 | 688 | |||
BC143341.1 | - | 6 | 674 | 9051846 |
