siRNA Details
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243116.1 | NP_001230045.1 | 2 | 83 | ||
NM_002503.4 | NP_002494.2 | 2 | 325 | |||
GenBank mRNA | AB209784.1 | - | 2 | 291 | ||
AB362577.1 | - | 2 | 81 | |||
AK290569.1 | - | 2 | 276 | |||
BC015528.1 | - | 2 | 256 | 3903794 | ||
BI520169.1 | - | 2 | 83 | |||
BM008311.1 | - | 2 | 277 | |||
BM449613.1 | - | 2 | 280 | |||
BT006743.1 | - | 2 | 222 | |||
DA633677.1 | - | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
