siRNA Details
Gene Symbol: CST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003650.3 | NP_003641.3 | 2 | 389 | ||
GenBank mRNA | AB015225.1 | - | 2 | 261 | ||
AF031824.1 | - | 2 | 328 | |||
AF036342.1 | - | 2 | 170 | |||
BC015507.1 | - | 2 | 284 | 3893020 | ||
BG547285.1 | - | 2 | 390 | |||
BT009825.1 | - | 2 | 179 | |||
CR541860.1 | - | 2 | 113 | |||
CR541878.1 | - | 2 | 113 |
