siRNA Details
Gene Symbol: MAPK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002749.3 | NP_002740.2 | 3 | 423 | ||
NM_139033.2 | NP_620602.2 | 3 | 634 | |||
NM_139034.2 | NP_620603.2 | 3 | 335 | |||
XM_006721557.3 | XP_006721620.1 | 3 | 381 | |||
XM_006721558.3 | XP_006721621.1 | 3 | 634 | |||
XM_006721559.3 | XP_006721622.1 | 3 | 543 | |||
GenBank mRNA | AB209611.1 | - | 1 | 1269 | ||
AK295376.1 | - | 3 | 328 | |||
AY534741.1 | - | 2 | 261 | |||
BC009963.2 | - | 3 | 355 | 4300124 | ||
BC030134.1 | - | 3 | 270 | 4111084 | ||
BP871947.1 | - | 3 | 330 | |||
DA452383.1 | - | 3 | 423 | |||
U25278.1 | - | 3 | 328 | |||
U29725.1 | - | 3 | 469 | |||
U29726.1 | - | 3 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
