siRNA Details
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 10 | 1852 | ||
NM_001284343.1 | NP_001271272.1 | 6 | 930 | |||
NM_007270.4 | NP_009201.2 | 9 | 1693 | |||
GenBank mRNA | AB209352.1 | - | 3 | 1077 | ||
AF089745.1 | - | 9 | 1364 | |||
AK075331.1 | - | 9 | 1657 | |||
AK128597.1 | - | 10 | 1634 | |||
AK293440.1 | - | 6 | 908 | |||
AK293954.1 | - | 8 | 1266 | |||
AK294266.1 | - | 8 | 1477 | |||
AK300328.1 | - | 10 | 1815 | |||
AK304643.1 | - | 6 | 768 | |||
AK316290.1 | - | 4 | 583 | |||
AL050187.1 | - | 1 | 23 | |||
BC007443.2 | - | 3 | 409 | |||
BC064418.1 | - | 4 | 616 | |||
BC072422.1 | - | 3 | 360 | |||
BC099918.1 | - | 2 | 255 | |||
BC101723.1 | - | 9 | 1621 | 8069229 | ||
BC112053.1 | - | 9 | 1621 | 8327521 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 5 | 672 | |||
NR_027339.1 | 3 | 776 | ||||
NR_027340.1 | 2 | 405 | ||||
NR_027342.1 | 3 | 686 | ||||
GenBank mRNA | AK289438.1 | - | 2 | 361 | ||
AK312981.1 | - | 2 | 405 | |||
BC011872.2 | - | 3 | 686 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
