siRNA Details
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000497.3 | NP_000488.3 | 8, 9 | 1410 | ||
XM_011516874.2 | XP_011515176.1 | 9, 10 | 1507 | |||
GenBank mRNA | AF478474.1 | - | 8, 9 | 1410 | ||
BC096285.3 | - | 10, 11 | 1625 | 40007097 | ||
BC096287.3 | - | 8, 9 | 1410 | 40007100 | ||
BX647738.1 | - | 3, 4 | 437 | |||
X55764.1 | - | 8, 9 | 1407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
