siRNA Details
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 5 | 904 | ||
NM_013421.2 | NP_038265.2 | 6 | 832 | |||
NM_013430.2 | NP_038347.2 | 5 | 592 | |||
GenBank mRNA | AL832738.1 | - | 808 | |||
BC025927.1 | - | 3 | 472 | 4548861 | ||
BU157907.1 | - | 5 | 575 | |||
BU164348.1 | - | 5 | 599 | |||
CR456494.1 | - | 2 | 182 | |||
J04131.1 | - | 6 | 774 | |||
J05235.1 | - | 4 | 849 | |||
M24087.1 | - | 1 | 107 | |||
M24903.1 | - | 5 | 592 | |||
X60069.1 | - | 3 | 463 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724392.3 | XP_006724455.1 | 4 | 655 | ||
XM_017029179.1 | XP_016884668.1 | 5 | 1336 | |||
XM_017029180.1 | XP_016884669.1 | 4 | 794 | |||
XM_017029181.1 | XP_016884670.1 | 4 | 1232 | |||
XM_017029182.1 | XP_016884671.1 | 5 | 1336 | |||
XM_017029185.1 | XP_016884674.1 | 5 | 816 | |||
XM_017029186.1 | XP_016884675.1 | 7 | 1495 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 2 | 301 | |||
GenBank mRNA | BC143607.1 | - | 2 | 167 |
Gene Symbol: GGT8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003503.1 | 1 | 707 | |||
GenBank mRNA | AK055838.1 | - | 1 | 707 |
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 3 | 940 | ||
XM_006724924.3 | XP_006724987.1 | 3 | 940 | |||
XM_011546256.2 | XP_011544558.1 | 3 | 943 | |||
XM_017030133.1 | XP_016885622.1 | 4 | 505 | |||
XM_017030134.1 | XP_016885623.1 | 4 | 508 | |||
XM_017030135.1 | XP_016885624.1 | 4 | 933 | |||
XM_017030136.1 | XP_016885625.1 | 5 | 975 | |||
XM_017030137.1 | XP_016885626.1 | 5 | 972 | |||
XM_017030138.1 | XP_016885627.1 | 5 | 495 | |||
XM_017030139.1 | XP_016885628.1 | 5 | 498 | |||
XM_017030140.1 | XP_016885629.1 | 6 | 965 | |||
XM_017030141.1 | XP_016885630.1 | 6 | 962 | |||
XM_017030142.1 | XP_016885631.1 | 4 | 806 | |||
XM_017030143.1 | XP_016885632.1 | 3 | 941 | |||
XM_017030145.1 | XP_016885634.1 | 3 | 940 | |||
XM_017030146.1 | XP_016885635.1 | 3 | 957 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 4 | 974 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
