siRNA Details
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288833.1 | NP_001275762.1 | 10 | 1555 | ||
NM_013421.2 | NP_038265.2 | 11 | 1483 | |||
NM_013430.2 | NP_038347.2 | 10 | 1243 | |||
GenBank mRNA | AL832738.1 | - | 1459 | |||
BC025927.1 | - | 8 | 1123 | 4548861 | ||
CR456494.1 | - | 7 | 833 | |||
J04131.1 | - | 11 | 1425 | |||
J05235.1 | - | 9 | 1500 | |||
M24087.1 | - | 6 | 758 | |||
M24903.1 | - | 10 | 1243 | |||
X60069.1 | - | 8 | 1114 |
Gene Symbol: GGT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005261890.4 | XP_005261947.1 | 3 | 309 | ||
XM_006724392.3 | XP_006724455.1 | 7 | 1017 | |||
XM_006724395.3 | XP_006724458.1 | 4 | 469 | |||
XM_017029179.1 | XP_016884668.1 | 8 | 1698 | |||
XM_017029180.1 | XP_016884669.1 | 7 | 1156 | |||
XM_017029181.1 | XP_016884670.1 | 7 | 1594 | |||
XM_017029182.1 | XP_016884671.1 | 8 | 1698 | |||
XM_017029183.1 | XP_016884672.1 | 3 | 285 | |||
XM_017029185.1 | XP_016884674.1 | 9 | 2538 | |||
XM_017029186.1 | XP_016884675.1 | 12 | 2161 |
Gene Symbol: GGT3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003267.1 | 7 | 950 | |||
GenBank mRNA | BC143607.1 | - | 4 | 442 |
Gene Symbol: LOC102724197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724921.3 | XP_006724984.1 | 7 | 1460 | ||
XM_006724924.3 | XP_006724987.1 | 6 | 1302 | |||
XM_006724928.3 | XP_006724991.1 | 2 | 498 | |||
XM_011546256.2 | XP_011544558.1 | 8 | 1594 | |||
XM_017030133.1 | XP_016885622.1 | 8 | 1025 | |||
XM_017030134.1 | XP_016885623.1 | 8 | 1028 | |||
XM_017030135.1 | XP_016885624.1 | 8 | 1453 | |||
XM_017030136.1 | XP_016885625.1 | 9 | 1495 | |||
XM_017030137.1 | XP_016885626.1 | 9 | 1492 | |||
XM_017030138.1 | XP_016885627.1 | 9 | 1015 | |||
XM_017030139.1 | XP_016885628.1 | 9 | 1018 | |||
XM_017030140.1 | XP_016885629.1 | 10 | 1485 | |||
XM_017030141.1 | XP_016885630.1 | 10 | 1482 | |||
XM_017030142.1 | XP_016885631.1 | 8 | 1326 | |||
XM_017030143.1 | XP_016885632.1 | 7 | 1461 | |||
XM_017030144.1 | XP_016885633.1 | 3 | 389 | |||
XM_017030145.1 | XP_016885634.1 | 4 | 1022 | |||
XM_017030146.1 | XP_016885635.1 | 7 | 1477 |
Gene Symbol: LOC102724823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030162.1 | XP_016885651.1 | 8 | 1494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
