siRNA Details
Gene Symbol: FOLH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153696.2 | NP_710163.1 | 4, 5 | 700 | ||
XM_017017319.1 | XP_016872808.1 | 9, 10 | 1053 | |||
GenBank mRNA | AF261715.1 | - | 4, 5 | 700 | ||
AY544126.1 | - | 3, 4 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
