siRNA Details
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001072.3 | NP_001063.2 | 1, 2 | 1008 | ||
NM_205862.1 | NP_995584.1 | 2, 3 | 221 | |||
GenBank mRNA | AY435141.1 | - | 1, 2 | 869 | ||
BC019861.1 | - | 1, 2 | 903 | 4454322 | ||
BM688740.1 | - | 2, 3 | 221 | |||
BM924331.1 | - | 2, 3 | 239 | |||
DQ364250.1 | - | 1, 2 | 877 | |||
J04093.1 | - | 1, 2 | 953 |
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000463.2 | NP_000454.1 | 1, 2 | 887 | ||
GenBank mRNA | AK290834.1 | - | 1, 2 | 891 | ||
AY435136.1 | - | 1, 2 | 872 | |||
BC128414.1 | - | 1, 2 | 888 | 40117598 | ||
BC128415.1 | - | 1, 2 | 887 | 40117599 | ||
DQ364247.1 | - | 1, 2 | 872 | |||
JQ686667.1 | - | 1, 2 | 872 | |||
JQ699637.1 | - | 1, 2 | 872 | |||
JQ699638.1 | - | 1, 2 | 872 | |||
JQ699639.1 | - | 1, 2 | 872 | |||
JQ699640.1 | - | 1, 2 | 872 | |||
JQ699641.1 | - | 1, 2 | 872 | |||
JQ699642.1 | - | 1, 2 | 872 | |||
JQ699643.1 | - | 1, 2 | 872 | |||
JQ699644.1 | - | 1, 2 | 872 | |||
JQ699645.1 | - | 1, 2 | 872 | |||
JQ699646.1 | - | 1, 2 | 872 | |||
JQ699647.1 | - | 1, 2 | 872 | |||
JQ699649.1 | - | 1, 2 | 872 | |||
JQ699650.1 | - | 1, 2 | 872 | |||
M57899.1 | - | 1, 2 | 887 |
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019093.2 | NP_061966.1 | 1, 2 | 875 | ||
GenBank mRNA | AY435138.1 | - | 1, 2 | 875 | ||
DQ364248.1 | - | 1, 2 | 143 |
Gene Symbol: UGT1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007120.2 | NP_009051.1 | 1, 2 | 904 | ||
GenBank mRNA | AK313623.1 | - | 1, 2 | 918 | ||
AY435139.1 | - | 1, 2 | 875 | |||
BC121036.1 | - | 2, 3 | 875 | 40117585 | ||
BC131623.1 | - | 1, 2 | 868 | |||
BC139784.1 | - | 1, 2 | 895 | 40117089 | ||
DQ364249.1 | - | 1, 2 | 884 | |||
M57951.1 | - | 1, 2 | 904 |
Gene Symbol: UGT1A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019078.1 | NP_061951.1 | 1, 2 | 875 | ||
GenBank mRNA | AY435140.1 | - | 1, 2 | 875 | ||
DQ383514.1 | - | 1, 2 | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
