siRNA Details
Gene Symbol: ALPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001632.4 | NP_001623.3 | 2 | 358 | ||
GenBank mRNA | AK075432.1 | - | 2 | 173 | ||
AK312931.1 | - | 2 | 358 | |||
BC009647.1 | - | 2 | 155 | 3460735 | ||
BC068501.1 | - | 2 | 141 | 30349481 | ||
DA851165.1 | - | 2 | 254 | |||
M12551.1 | - | 2 | 99 | |||
M14169.1 | - | 1 | 38 | |||
M14170.1 | - | 2 | 103 |
Gene Symbol: ALPPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031313.2 | NP_112603.2 | 2 | 133 | ||
GenBank mRNA | AK293017.1 | - | 2 | 131 | ||
BC014139.2 | - | 2 | 118 | 4652865 | ||
DB194270.1 | - | 2 | 133 | |||
J04948.1 | - | 2 | 132 | |||
X53279.1 | - | 2 | 83 | |||
X55958.1 | - | 2 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
