siRNA Details
Gene Symbol: TMEM189-UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199203.2 | NP_954673.1 | 4 | 554 | ||
GenBank mRNA | BF982644.1 | - | 3 | 201 | ||
BG723381.1 | - | 4 | 484 | |||
BI838424.1 | - | 4 | 470 | |||
U39361.1 | - | 1 | 15 |
Gene Symbol: TMEM189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162505.1 | NP_001155977.1 | 4 | 545 | ||
NM_199129.2 | NP_954580.1 | 4 | 554 | |||
NR_027889.1 | 5 | 658 | ||||
GenBank mRNA | AB044550.1 | - | 4 | 467 | ||
AF155120.1 | - | 4 | 481 | |||
AK023028.1 | - | 4 | 479 | |||
AK304595.1 | - | 3 | 269 | |||
BC018893.2 | - | 4 | 466 | |||
BC142966.1 | - | 4 | 469 | 8860386 | ||
BC144612.1 | - | 5 | 658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
