siRNA Details
Gene Symbol: TPTEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001591.1 | 1, 2 | 303 | |||
GenBank mRNA | AY219889.1 | - | 1, 2 | 303 | ||
EU541207.1 | - | 1, 2 | 268 | |||
EU541208.1 | - | 1, 2 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
