siRNA Details
Gene Symbol: NEBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006393.2 | NP_006384.1 | 2 | 498 | ||
XM_005252342.4 | XP_005252399.1 | 2 | 602 | |||
XM_005252343.4 | XP_005252400.1 | 2 | 602 | |||
XM_005252344.4 | XP_005252401.1 | 2 | 602 | |||
XM_011519291.2 | XP_011517593.1 | 4 | 494 | |||
XM_017015467.1 | XP_016870956.1 | 4 | 610 | |||
XM_017015468.1 | XP_016870957.1 | 5 | 378 | |||
XM_017015469.1 | XP_016870958.1 | 2 | 602 | |||
GenBank mRNA | BC110452.1 | - | 2 | 185 | 40027448 | |
BC110453.1 | - | 2 | 185 | 40027449 | ||
Y16241.1 | - | 2 | 498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
