siRNA Details
Gene Symbol: ZBTB44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301098.1 | NP_001288027.1 | 2 | 951 | ||
NM_001301099.1 | NP_001288028.1 | 2 | 951 | |||
NM_014155.4 | NP_054874.3 | 2 | 951 | |||
XM_005271521.4 | XP_005271578.1 | 2 | 1147 | |||
XM_005271522.4 | XP_005271579.1 | 2 | 1026 | |||
XM_006718825.3 | XP_006718888.1 | 2 | 1026 | |||
XM_006718826.3 | XP_006718889.1 | 2 | 1023 | |||
XM_011542789.2 | XP_011541091.1 | 2 | 1024 | |||
XM_017017623.1 | XP_016873112.1 | 2 | 1023 | |||
XM_017017624.1 | XP_016873113.1 | 2 | 1023 | |||
GenBank mRNA | AF161548.1 | - | 2 | 670 | ||
AK024659.1 | - | 1 | 118 | |||
AK314701.1 | - | 2 | 828 | |||
BC030580.1 | - | 2 | 835 | 4823171 | ||
BC050723.1 | - | 1 | 573 | 4445874 | ||
BC071729.1 | - | 2 | 930 | 5163748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
