siRNA Details
Gene Symbol: RABGAP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014857.4 | NP_055672.3 | 2, 3 | 412 | ||
XM_005245680.1 | XP_005245737.1 | 2, 3 | 342 | |||
XM_006711693.1 | XP_006711756.1 | 2, 3 | 342 | |||
XM_011510223.2 | XP_011508525.1 | 3, 4 | 369 | |||
GenBank mRNA | AB449904.1 | - | 1, 2 | 135 | ||
AK095838.2 | - | 2, 3 | 309 | |||
AL157958.1 | - | 2, 3 | 236 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
