siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 629 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 626 | |||
NR_073500.1 | 2 | 324 | ||||
NR_073501.1 | 5 | 700 | ||||
NR_073502.1 | 3 | 579 | ||||
NR_073503.1 | 3 | 576 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 367 | ||
AB097043.1 | - | 3 | 367 | |||
AB097044.1 | - | 3 | 358 | |||
AB097045.1 | - | 3 | 358 | |||
AB097046.1 | - | 4 | 417 | |||
AJ295987.1 | - | 3 | 363 | |||
AK297926.1 | - | 4 | 417 | |||
AK308787.1 | - | 2 | 111 | |||
AL050028.1 | - | 3 | 363 | |||
BC023988.1 | - | 3 | 384 | 3825796 | ||
BC065538.1 | - | 3 | 364 | 6053254 | ||
DA010145.1 | - | 4 | 421 | |||
DA438597.1 | - | 5 | 487 |
Gene Symbol: LOC100130075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073494.1 | 1 | 433 |
