siRNA Details
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 1265 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 1262 | |||
NR_073500.1 | 2 | 960 | ||||
NR_073501.1 | 5 | 1336 | ||||
NR_073502.1 | 3 | 1215 | ||||
NR_073503.1 | 3 | 1212 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 1009 | ||
AB097043.1 | - | 3 | 1009 | |||
AB097046.1 | - | 4 | 1053 | |||
AJ295987.1 | - | 3 | 999 | |||
AK298576.1 | - | 4 | 950 | |||
AK308787.1 | - | 2 | 747 | |||
AK316205.1 | - | 4 | 939 | |||
AL050028.1 | - | 3 | 999 | |||
BC023988.1 | - | 3 | 1020 | 3825796 | ||
BX640985.1 | - | 3 | 611 |
