siRNA Details
Gene Symbol: REG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF172331.1 | - | 5 | 561 | ||
AK291981.1 | - | 6 | 582 | |||
BC005350.2 | - | 5 | 558 | 3950349 | ||
BM769434.1 | - | 6 | 625 | |||
M18963.1 | - | 5 | 566 | |||
M27190.1 | - | 5 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
