siRNA Details
Gene Symbol: TMEM138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016464.4 | NP_057548.1 | 5 | 1198 | ||
NR_028473.1 | 6 | 1081 | ||||
XM_006718585.3 | XP_006718648.1 | 4 | 1436 | |||
XM_006718586.2 | XP_006718649.1 | 5 | 835 | |||
XM_006718588.3 | XP_006718651.1 | 5 | 693 | |||
GenBank mRNA | AF151030.1 | - | 5 | 815 | ||
AF151032.1 | - | 5 | 812 | |||
AL832879.1 | - | 3 | 3351 | |||
BC005201.1 | - | 5 | 813 | 3684390 | ||
CB956757.1 | - | 6 | 754 |
