siRNA Details
Gene Symbol: CEND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016564.3 | NP_057648.2 | 2 | 1381 | ||
GenBank mRNA | AF235030.1 | - | 1 | 1287 | ||
AK074547.1 | - | 2 | 1377 | |||
BC034732.1 | - | 2 | 1367 | 5179740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
